Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: killer cell lectin-like receptor subfamily A, member 9; Ly-49; Ly-49I receptor; Ly49I receptor
Gene Aliases: Ly49I; LY49I1
Entrez Gene ID: (Mouse) 16640
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support