Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: SLENTLTESE AHYSSQLSQV QSLITNVESQ LAEIRCDLER QNQEYQVLLD VRARLECEIN TYRSLLESED CKLPCNPCAT TNASGNSCGP CGTSQKGCCN
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hair keratin; Hair keratin, type I Ha4; hard keratin, type I, 4; K34; keratin 34, type I; keratin, hair, acidic, 4; Keratin, type I cuticular Ha4; Keratin-34; KRT hHa4
Gene Aliases: Ha-4; HA4; HHA4; HKA4; KRT34; KRTHA4
UniProt ID: (Human) O76011
Entrez Gene ID: (Human) 3885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support