Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: YHLMPERRP HLPAFRTRPR CCTSIAGLIY AVGGLNSAGD SLNVVEVFDP IANCWERCRP MTTARSRVGV AVVNGLLYAI GGYDGQLRLS TVEAYNPETD TWTRVGSMNS KRSAMGTVVL DGQIYVCGGY DGNSSLSSVE TYSPETDKWT VVTSMSSNRS AAGVTVFEGR IYVSGGHDGL QIFSSVEHYN HHTATWHPAA GMLNKRCRHG AASLGSKMFV CGGYDGSGFL SIAEMYSSVA DQWCLIVPMH TRRSRVSLVA SCGRLYAVGG YDGQSNLSSV EMYDPETDCW TFMAPMACHE GGVGVGCIPL LTI (198-509 aa encoded by BC032620)
KLHL18 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like 18; kelch-like family member 18; Kelch-like protein 18
Gene Aliases: A930041K15; AW545966; KIAA0795; KLHL18; OK/SW-cl.74; RGD1310570
UniProt ID: (Human) O94889, (Mouse) E9Q4F2
Entrez Gene ID: (Human) 23276, (Rat) 316012, (Mouse) 270201
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support