Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SAAGVTVFEG RIYVSGGHDG LQIFSSVEHY NHHTATWHPA AGMLNKRCRH GAASLGSKMF VCGGYDGSGF LSIAEMYSSV ADQWCLIVPM HT
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
KLHL18 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like family member 18; Kelch-like protein 18
Gene Aliases: KIAA0795; KLHL18; OK/SW-cl.74
UniProt ID: (Human) O94889
Entrez Gene ID: (Human) 23276
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support