Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MGKTNSKLAP EVLEDLVQNT EFSEQELKQW YKGFLKDCPS GILNLEEFQQ LYIKFFPYGD ASKFAQHAFR TFDKNGDGTI DFREFICALS VTSRGSFEQK LNWAFEMYDL DGDGRITRLE MLEIIEAIYK MVGTVIMMRM NQDGLTPQQR VDKIFKKMDQ DKDDQITLEE FKEAAKSDAS IVLLLQCDMQ K
May be involved in the calcium-dependent regulation of rhodopsin phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hippocalcin-like protein 4; HLP4
Gene Aliases: HLP4; HPCAL4
UniProt ID: (Human) Q9UM19
Entrez Gene ID: (Human) 51440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support