Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
May play an important role in the development of cancers in a broad range of tissues. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Down-regulated in multiple cancers 1; downregulated in multiple cancer 1; HID1 domain containing; HID1 domain-containing protein; Protein hid-1 homolog; Protein HID1; UPF0663 transmembrane protein C17orf28
Gene Aliases: C17orf28; C9H17orf28; DMC1; HID-1; HID1; RGD1311422
UniProt ID: (Human) Q8IV36
Entrez Gene ID: (Dog) 483305, (Human) 283987, (Rat) 287822
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support