PA1-20524 detects G Protein-Coupled Receptor GPR81 in human samples.
PA1-20524 has been successfully used in immunohistochemistry (Paraffin) procedures.
The PA1-20524 immunogen is a synthetic peptide corresponding to the C-terminal portion of human G Protein-Coupled Receptor GPR81.
The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.