Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MIQKGCTPSG EGADSEPRAE TVPEEANYEA VPVEAYGLAM LRGMGWKPGE GIGRTFNQVV KPRVNSLRPK GLGLGANLTE AQALTPTGPS RMPRPDEEQE KDKEDQPQGL VPGGAVVVLS GPHRGLYGKV EGLDPDNVRA MVRLAVGSRV VTVSEYYLRP VSQQEFDKNT LDLRQQNGTA SSRKTLWNQE LYIQQDNSER KRKHLPDRQD GPAAKSEKAA PRSQHWLHRD LRVRFVDNMY KGGQYYNTKM IIEDVLSPDT CVCRTDEGRV LEGLREDMLE TLVPKAEGDR VMVVLGPQTG RVGHLLSRDR ARSRALVQLP RENQVVELHY DAICQYMGPS DTDDD
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G patch domain and KOW motifs; G patch domain and KOW motifs-containing protein; G-patch domain and KOW motifs-containing protein; G-patch domain-containing protein 5; Protein MOS2 homolog; Protein T54
Gene Aliases: 4432411B18; DXErtd697e; DXImx41e; GPATC5; GPATCH5; GPKOW; Spp2; T54
UniProt ID: (Human) Q92917, (Mouse) Q56A08
Entrez Gene ID: (Human) 27238, (Rat) 679890, (Mouse) 209416
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support