Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTSANKAIEL QLQVKQNAEE LQDFMRDLEN WEKDIKQKDM ELRRQNGVPE ENLPPIRNGN FRKKKKGKAK ESSKKTREEN TKNRIKSYDY EAWAKLDVDR ILDELDKDDS THESLSQESE SEEDGIHVDS QKALVLKEKG NKYFKQGKYD EAIDCYTKGM DADPYNPVLP TNRASAYFRL KKFAVAESDC NLAVALNRSY TKAYSRRGAA RFALQKLEEA KKDYERVLEL EPNNFEATNE LRKISQALAS KENSYPKEAD IVIKSTEGER KQIEAQQNKQ QAISEKDRGN GFFKEGKYER AIECYTRGIA ADGANALLPA NRAMAYLKIQ KYEEAEKDCT QAILLDGSYS KAFARRGTAR TFLGKLNEAK QDFETVLLLE PGNKQAVTEL SKIKKKPLKK VIIEETGNLI QTIDVPDSTT AAAPENNPIN LANVIAATGT TSKKNSSQDD LFPTSDTPRA KVLKIEEVSD TSSLQPQASL KQDVCQSYSE KMPIEIEQKP AQFATTVLPP IPANSFQLES DFRQLKSSPD MLYQYLKQIE PSLYPKLFQK NLDPDVFNQI VKILHDFYIE KEKPLLIFEI LQRLSELKRF DMAVMFMSET EKKIARALFN HIDKSGLKDS SVEELKKRYG G
This gene encodes an RNA polymerase II-associated protein. The encoded protein may function in transcriptional regulation and may also regulate apoptosis. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase II-associated protein 3
Gene Aliases: RPAP3
UniProt ID: (Human) Q9H6T3
Entrez Gene ID: (Human) 79657
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support