Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DGVGNSYIE DNDDDSKMAD LLSYFQQQLT FQESVLKLCQ PELESSQIHI SVLPMEVLMY IFRWVVSSDL DLRSLEQLSL VCRGFYICAR DPEIWRLACL KVWGRSCIKL VPYTSWREMF LERPRVRFDG VYISKTTYIR QGEQSLDGFY RAWHQVEYYR YIRFFPDGHV MMLTTPEEPQ SIVPRLRTRN TRTDAILLGH YRLSQDTDNQ TKVFAVITKK KEEKPLDYKY RYFRRVPVQE ADQSFHVGLQ LCSSGHQRFN KLIWIHHSCH ITYKSTGETA VSAFEIDKMY TPLFFARVRS YTAFSERPL (140-447 aa encoded by BC000650)
FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cross-immune reaction antigen 1; F-box only protein 9; F-box protein Fbx9; Renal carcinoma antigen NY-REN-57
Gene Aliases: 9030401P18Rik; AA986398; dJ341E18.2; FBX9; FBXO9; MNCb-2471; NY-REN-57; VCIA1
UniProt ID: (Human) Q9UK97, (Mouse) Q8VDY6, (Rat) Q5U2X1
Entrez Gene ID: (Human) 26268, (Mouse) 71538, (Rat) 300849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support