Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Human CD41 antibody, clone PM6/248 recognizes the human CD41 cell surface antigen, a approximately 140 kDa glycoprotein expressed by platelets and megakaryocytes.
CD41, also known as platelet glycoprotein IIb or ITGA2B, is a protein composed of two subunits: a 120 kDa alpha subunit and a 23 kDa beta subunit. It interacts with CD61 (gpIIIa, integrin beta III) in the presence of calcium to form a functional adhesive protein receptor. Initially thought to be expressed exclusively on platelets and megakaryocytes, CD41 is also found on hematopoietic progenitors in the embryo, fetus, and adult, indicating its role in early stages of hematopoietic differentiation. CD41 plays a crucial role in platelet function and blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to various adhesion molecules, including von Willebrand factor, fibrinogen, fibronectin, and vitronectin, facilitating platelet adhesion and aggregation. This receptor is essential for platelet function, contributing to the binding of several adhesion molecules and playing a significant role in hemostasis. Diseases associated with CD41 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 bleeding disorder, highlighting its importance in normal platelet function and blood coagulation processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: alphaIIb protein; CD41; form 1; form 2; GPalpha IIb; GPIIb; Integrin alpha-IIb; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb of IIb/IIIa complex; Platelet membrane glycoprotein IIb; platelet-specific antigen BAK; protein phosphatase 1, regulatory subunit 93
Gene Aliases: BDPLT16; BDPLT2; CD41; CD41B; GP2B; GPIIb; GT; GTA; HPA3; ITGA2B; ITGAB; PPP1R93
UniProt ID: (Human) P08514
Entrez Gene ID: (Human) 3674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support