Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: SASLSNLHSL DRSTREVELG LEYGSPTMNL AGQSLKFENG QWIAETGVSG GVDRREVQRL RRRNQQLEEE NNLLRLKVDI LLDMLSESTA ESHLMEKELD ELRISRKRK
Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARPP-binding protein; chibby CTNNB1-mediated transcription inhibitor; chibby homolog 1; coiled-coil protein PIGEA-14; Cytosolic leucine-rich protein; PIGEA-14; PKD2 interactor, Golgi and endoplasmic reticulum-associated 1; polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, 14 kDa; Protein chibby homolog 1
Gene Aliases: ARB1; C22orf2; CBY; CBY1; HRIHFB2025; HS508I15A; PGEA1; PIGEA-14; PIGEA14
UniProt ID: (Human) Q9Y3M2
Entrez Gene ID: (Human) 25776
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support