Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The MW of this protein is 34 kDa, and this antibody specifically recognizes the 34 kDa protein. This antibody works well in WB, IHC and IF applications.
Immunogen sequence: MPVVRKIFR RRRGDSESEE DEQDSEEVRL KLEETREVQN LRKRPNGVSA VALLVGEKVQ EETTLVDDPF QMKTGGMVDM KKLKERGKDK ISEEEDLHLG TSFSAETNRR DEDADMMKYI ETELKKRKGI VEHEEQKVKP KNAEDCLYEL PENIRVSSAK KTEEMLSNQM LSGIPEVDLG IDAKIKNIIS TEDAKARLLA EQQNKKKDSE TSFVPTNMAV NYVQHNRFYH EELNAPIRRN KEEPKARPLR VGDTEKPEPE RSPPNRKRPA NEKATDDYHY EKFKKMNRRY (1-289 aa encoded by BC007664)
Involved in the regulation of telomeric heterochromatin assembly and control of telomere length. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma-associated antigen 59; Splicing factor C9orf78; uncharacterized protein C9orf78 homolog
Gene Aliases: bA409K20.3; C9orf78; HCA59; HSPC220
UniProt ID: (Human) Q9NZ63
Entrez Gene ID: (Human) 51759, (Rat) 311855
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support