Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTPSEGARA GTGRELEMLD SLLALGGLVL LRDSVEWEGR SLLKALVKKS ALCGEQVHIL GCEVSEEEFR EGFDSDINNR LVYHDFFRDP LNWSKTEEAF PGGPLGALRA MCKRTDPVPV TIALDSLSWL LLRLPCTTLC QVLHAVSHQD SCPGDSSSVG KVSVLGLLHE ELHGPGPVGA LSSLAQTEVT LGGTMGQASA HILCRRPRQR (1-209 aa encoded by BC002762)
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dermal papilla derived protein 6; Dermal papilla-derived protein 6; Elongator complex protein 5; S-phase 2 protein
Gene Aliases: C17orf81; DERP6; ELP5; HSPC002; MST071; MSTP071
UniProt ID: (Human) Q8TE02
Entrez Gene ID: (Human) 23587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support