Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MGSTDSKLN FRKAVIQLTT KTQPVEATDD AFWDQFWADT ATSVQDVFAL VPAAEIRAVR EESPSNLATL CYKAVEKLVQ GAESGCHSEK EKQIVLNCSR LLTRVLPYIF EDPDWRGFFW STVPGAGRGG QGEEDDEHAR PLAESLLLAI ADLLFCPDFT VQSHRRSTVD SAEDVHSLDS CEYIWEAGVG FAHSPQPNYI HDMNRMELLK LLLTCFSEAM YLPPAPESGS TNPWVQFFCS TENRHALPLF TSLLNTVCAY DPVGYGIPYN HLLFSDYREP LVEEAAQVLI VTLDHDSASS ASPTVDGTTT GTAMDDADPP GPENLFVNYL SRIHREEDFQ FILKGIARLL S (1-350 aa encoded by BC035372)
May play an important role in the development of cancers in a broad range of tissues. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Down-regulated in multiple cancers 1; downregulated in multiple cancer 1; HID1 domain containing; HID1 domain-containing protein; Protein hid-1 homolog; Protein HID1; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Gene Aliases: C17orf28; C630004H02Rik; DMC1; HID-1; HID1; RGD1311422
UniProt ID: (Human) Q8IV36, (Mouse) Q8R1F6
Entrez Gene ID: (Human) 283987, (Rat) 287822, (Mouse) 217310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support