Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGAAPTTAF GQAVIGPPGS GKTTYCLGMS EFLRALGRRV AVVNLDPANE GLPYECAVDV GELVGLGDVM DALRLGPNGG LLYCMEYLEA NLDWLRAKLD PLRGHYFLFD CPGQVELCTH HGALRSIFSQ MAQWDLRLTA VHLVDSHYCT DPAKFISVLC TSLATMLHVE LPHINLLSKM DLIEHYGKLA FNLDYYTEVL DLSYLLDHLA SDPFFRHYRQ LNEKLVRLIE DYSLVSFIPL NIQDKESIQR VLQAVDKANG YCFGAQEQRS LEAMMSAAMG ADFHFSSTLG IQEKYLAPSN QSVEQEAMQL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP binding domain 1 family, member B; ATP-binding domain 1 family member B; ATPBD1B; GPN-loop GTPase 2
Gene Aliases: ATPBD1B; GPN2; UNQ5828/PRO19647
UniProt ID: (Human) Q9NW30
Entrez Gene ID: (Human) 54707
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support