Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SIELVEAEV SELETRLEKL LKLGTGLLES GRHYLAASRA FVVGICDLAR LGPPEPMMAE CLEKFTVSLN HKLDSHAELL DATQHTLQQQ IQTLVKEGLR GFREARRDFW RGAESLEAAL THNAEVPRRR AQEAEEAGAA LRTARAGYRG RALDYALQIN VIEDKRKFDI MEFVLRLVEA QATHFQQGHE ELSRLSQYRK ELGAQLHQLV LNSAREKRDM EQRHVLLKQK ELG (20-251 aa encoded by BC018543)
ACAP1, also known as Centaurin-beta-1 (CENTB1), is a 740 amino acid containing transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain and a PH domain that binds to phospholipids. ACAP1, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6), is required for clathrin-dependent export of proteins from recycling endosomes to trans-Golgi network and cell surface. It mediates CENTB1-binding to phosphatidylinositol 3,5-bisphosphate (PIP2) or phosphatidylinositol 3,4,5-trisphosphate (PIP3) containing membranes. Along with ACAP2, ACAP1 is recruited to platelet-derived growth factor (PDGF) induced dorsal membrane ruffles in NIH 3T3 mouse fibroblasts, while over expression inhibited ruffle formation. It is widely expressed in most tissues with high expression in lung and spleen and low expression in heart, kidney, liver and pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf GAP with coiled coil, ANK repeat and PH domains 1; Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1; centaurin beta 1; centaurin, beta 1; Centaurin-beta-1; Cnt-b1
Gene Aliases: ACAP1; CENTB1; KIAA0050
UniProt ID: (Human) Q15027
Entrez Gene ID: (Human) 9744
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support