Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: CPDICFRVAG CSFLCHKAFF CGRSDYFRAL LDDHFRESEE PATSGGPPAV TLHGISPDVF THVLYYMYSD HTELSPEAAY DVLSVADMYL LPGLKRLCGR
This gene encodes a protein with an ankyrin repeat region and two BTB/POZ domains, which are thought to be involved in protein-protein interactions. Expression of this gene is activated by the phosphatase and tensin homolog, a tumor suppressor. Alternate splicing results in three transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABTB1; ankyrin repeat and BTB (POZ) domain containing 1; Ankyrin repeat and BTB/POZ domain-containing protein 1; Elongation factor 1A-binding protein; MGC20585
Gene Aliases: ABTB1; BPOZ; BTB3; BTBD21; EF1ABP; PP2259
UniProt ID: (Human) Q969K4
Entrez Gene ID: (Human) 80325
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support